Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018830.1 | NP_061300.1 | 16 - 17 | 1892 | 69 | |
GenBank mRNA | AB037111.1 | - | 17 - 18 | 2177 | 69 | |
AB037181.1 | - | 16 - 17 | 1892 | 69 | ||
AK080951.1 | - | 16 - 17 | 2069 | 69 | ||
BC022604.1 | - | 7 - 8 | 674 | 69 | ||
BC137898.1 | - | 16 - 17 | 1875 | 8860917 | 69 | |
BC145016.1 | - | 16 - 17 | 1875 | 9053564 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |