Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133352.2 | NP_579930.1 | 6 - 7 | 934 | 132 | |
GenBank mRNA | AF269151.1 | - | 6 - 7 | 866 | 132 | |
AK075807.1 | - | 6 - 7 | 902 | 132 | ||
AK147380.1 | - | 6 - 7 | 929 | 132 | ||
AK147629.1 | - | 6 - 7 | 928 | 132 | ||
AK220279.1 | - | 6 - 7 | 910 | 132 | ||
BC004799.1 | - | 5 - 6 | 563 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |