Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293757.1 | NP_001280686.1 | 1 - 2 | 295 | 78 | |
NM_001293759.1 | NP_001280688.1 | 1 - 2 | 295 | 78 | ||
NM_011722.4 | NP_035852.1 | 1 - 2 | 295 | 78 | ||
GenBank mRNA | AF190796.1 | - | 1 - 2 | 36 | 78 | |
AK003618.1 | - | 1 - 2 | 35 | 78 | ||
BC029249.1 | - | 1 - 2 | 36 | 4486579 | 78 | |
BY079663.1 | - | 1 - 2 | 95 | 78 | ||
CA573939.1 | - | 1 - 2 | 295 | 78 | ||
DV062065.1 | - | 1 - 2 | 87 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |