Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205082.1 | NP_001192011.1 | 7 - 8 | 1228 | 88 | |
NM_001205083.1 | NP_001192012.1 | 7 - 8 | 1228 | 88 | ||
NM_016741.2 | NP_058021.1 | 7 - 8 | 1228 | 88 | ||
GenBank mRNA | AK010622.1 | - | 7 - 8 | 1153 | 88 | |
AK028191.1 | - | 7 - 8 | 1209 | 88 | ||
AK033114.1 | - | 7 - 8 | 1153 | 88 | ||
AK145588.1 | - | 7 - 8 | 1154 | 88 | ||
AK154933.1 | - | 7 - 8 | 1220 | 88 | ||
AK158122.1 | - | 7 - 8 | 1153 | 88 | ||
AK169906.1 | - | 7 - 8 | 1204 | 88 | ||
AK171602.1 | - | 7 - 8 | 1208 | 88 | ||
BC004656.1 | - | 7 - 8 | 1208 | 3498018 | 88 | |
CT010222.1 | - | 7 - 8 | 1016 | 88 | ||
U37799.1 | - | 7 - 8 | 1066 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |