Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037758.2 | NP_001032847.2 | 3 - 4 | 346 | 116 | |
NM_001286465.1 | NP_001273394.1 | 3 - 4 | 346 | 116 | ||
NM_001286466.1 | NP_001273395.1 | 3 - 4 | 346 | 116 | ||
NM_009771.3 | NP_033901.1 | 2 - 3 | 238 | 116 | ||
GenBank mRNA | AF081887.1 | - | 2 - 3 | 223 | 116 | |
AF099932.1 | - | 2 - 3 | 133 | 116 | ||
AF110396.1 | - | 2 - 3 | 133 | 116 | ||
AF112979.1 | - | 2 - 3 | 133 | 116 | ||
AK032221.1 | - | 3 - 4 | 309 | 116 | ||
AK052317.1 | - | 4 - 5 | 370 | 116 | ||
AK083257.1 | - | 3 - 4 | 117 | |||
AK145091.1 | - | 3 - 4 | 282 | 116 | ||
AK145624.1 | - | 3 - 4 | 329 | 116 | ||
AK156660.1 | - | 2 - 3 | 266 | 116 | ||
AK180045.1 | - | 3 - 4 | 225 | 116 | ||
AK220183.1 | - | 3 - 4 | 344 | 116 | ||
BC003989.1 | - | 2 - 3 | 139 | 3491843 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |