Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195265.1 | NP_001182194.1 | 6 - 7 | 1061 | 73 | |
GenBank mRNA | KF041446.1 | - | 6 - 7 | 932 | 73 | |
KF041447.1 | - | 6 - 7 | 932 | 73 | ||
KF041451.1 | - | 6 - 7 | 932 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |