Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289543.1 | NP_001276472.1 | 6 - 7 | 980 | 90 | |
NM_001289544.1 | NP_001276473.1 | 7 - 8 | 1304 | 90 | ||
NM_028227.3 | NP_082503.2 | 7 - 8 | 1176 | 90 | ||
GenBank mRNA | AF321920.1 | - | 7 - 8 | 1068 | 90 | |
AF321921.1 | - | 7 - 8 | 1305 | 90 | ||
AK013885.1 | - | 5 - 6 | 669 | 90 | ||
AK034212.1 | - | 2 - 3 | 109 | 90 | ||
AK143718.1 | - | 6 - 7 | 970 | 90 | ||
BC038490.1 | - | 4 - 5 | 545 | 3599718 | 90 | |
BC062932.1 | - | 7 - 8 | 1152 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |