Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164311.1 | NP_001157783.1 | 9 - 10 | 1539 | 64 | |
NM_053083.3 | NP_444313.3 | 9 - 10 | 1536 | 64 | ||
GenBank mRNA | AF338440.1 | - | 8 - 9 | 1431 | 64 | |
AK014331.1 | - | 9 - 10 | 1539 | 64 | ||
BC068307.1 | - | 9 - 10 | 1478 | 30059745 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |