Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500027922.mm.1 |
TC0500003138.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027922.mm.1 |
NM_001109902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027922.mm.1 |
NM_001109902.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027922.mm.1 |
NM_172998.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027922.mm.1 |
NM_172998.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027921.mm.1 |
TC0500003138.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027921.mm.1 |
NM_001109902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027921.mm.1 |
NM_001109902.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027921.mm.1 |
NM_172998.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027921.mm.1 |
NM_172998.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027930.mm.1 |
TC0500003138.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027930.mm.1 |
NM_001109902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027930.mm.1 |
NM_001109902.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027924.mm.1 |
TC0500003138.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027924.mm.1 |
NM_001109902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027924.mm.1 |
NM_001109902.1 |
Refseq |