Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109902.1 | NP_001103372.1 | 8 - 9 | 1245 | 63 | |
NM_172998.3 | NP_766586.2 | 9 - 10 | 1280 | 63 | ||
GenBank mRNA | AK046850.1 | - | 9 - 10 | 1281 | 63 | |
AK090256.1 | - | 8 - 9 | 1341 | 63 | ||
AK148930.1 | - | 8 - 9 | 1222 | 63 | ||
BC057302.1 | - | 8 - 9 | 1207 | 6827287 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |