Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019734.2 | NP_062708.1 | 3 - 4 | 330 | 134 | |
GenBank mRNA | AF157500.1 | - | 3 - 4 | 260 | 134 | |
AF352179.1 | - | 3 - 4 | 272 | 134 | ||
AK075666.1 | - | 3 - 4 | 245 | 134 | ||
AK075919.1 | - | 3 - 4 | 284 | 134 | ||
AK077721.1 | - | 3 - 4 | 283 | 134 | ||
AK090314.1 | - | 3 - 4 | 280 | 134 | ||
AK150567.1 | - | 3 - 4 | 243 | 134 | ||
AK151208.1 | - | 3 - 4 | 331 | 134 | ||
AK151312.1 | - | 3 - 4 | 243 | 134 | ||
AK151478.1 | - | 3 - 4 | 321 | 134 | ||
AK152305.1 | - | 3 - 4 | 282 | 134 | ||
AK152811.1 | - | 3 - 4 | 243 | 134 | ||
AK152920.1 | - | 3 - 4 | 290 | 134 | ||
AK153101.1 | - | 3 - 4 | 290 | 134 | ||
AK153297.1 | - | 3 - 4 | 244 | 134 | ||
AK159556.1 | - | 3 - 4 | 252 | 134 | ||
AK166757.1 | - | 3 - 4 | 262 | 134 | ||
BC003204.1 | - | 3 - 4 | 250 | 3587042 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |