Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145123.4 | NP_660105.3 | 10 - 11 | 1466 | 61 | |
GenBank mRNA | AJ421516.1 | - | 10 - 11 | 1316 | 61 | |
AK032328.1 | - | 10 - 11 | 1331 | 61 | ||
AK049801.1 | - | 10 - 11 | 1437 | 61 | ||
AK083221.1 | - | 10 - 11 | 1470 | 61 | ||
AK149020.1 | - | 10 - 11 | 1472 | 61 | ||
AK149069.1 | - | 10 - 11 | 1467 | 61 | ||
BC024472.1 | - | 7 - 8 | 902 | 4976509 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |