Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500009339.mm.1 |
TC0500001099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009339.mm.1 |
NM_001081234.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009341.mm.1 |
TC0500001099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009341.mm.1 |
NM_001081234.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009344.mm.1 |
TC0500001099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009344.mm.1 |
NM_001081234.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009340.mm.1 |
TC0500001099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009340.mm.1 |
NM_001081234.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009342.mm.1 |
TC0500001099.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009342.mm.1 |
NM_001081234.2 |
Refseq |