Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164362.1 | NP_001157834.1 | 3 - 4 | 630 | 98 | |
NM_028293.1 | NP_082569.1 | 2 - 3 | 254 | 98 | ||
NM_028760.2 | NP_083036.2 | 3 - 4 | 606 | 98 | ||
GenBank mRNA | AK004655.1 | - | 3 - 4 | 632 | 98 | |
AK012311.1 | - | 3 - 4 | 403 | 98 | ||
AK028216.1 | - | 2 - 3 | 240 | 98 | ||
CJ074252.1 | - | 2 - 3 | 257 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |