Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwf19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081077.1 | NP_001074546.1 | 1 - 2 | 116 | 128 | |
GenBank mRNA | AK139654.1 | - | 1 - 2 | 117 | 128 | |
BC027553.1 | - | 1 - 2 | 101 | 128 | ||
BC037640.1 | - | 1 - 2 | 94 | 5096937 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |