Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144846.5 | NP_659095.1 | 6 - 7 | 741 | 120 | |
GenBank mRNA | AK028062.1 | - | 6 - 7 | 742 | 120 | |
AK075613.1 | - | 7 - 8 | 686 | 120 | ||
AK215696.1 | - | 1 - 2 | 103 | 120 | ||
BC011343.1 | - | 6 - 7 | 559 | 4012217 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |