Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144846.5 | NP_659095.1 | 7 - 8 | 814 | 146 | |
GenBank mRNA | AK028062.1 | - | 7 - 8 | 815 | 146 | |
AK075613.1 | - | 8 - 9 | 759 | 146 | ||
BC011343.1 | - | 7 - 8 | 632 | 4012217 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |