Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053103.5 | NP_444333.3 | 4 - 5 | 708 | 56 | |
GenBank mRNA | AK007446.1 | - | 4 - 5 | 580 | 56 | |
AK081564.1 | - | 4 - 5 | 566 | 56 | ||
AK147401.1 | - | 4 - 5 | 573 | 56 | ||
AK166220.1 | - | 4 - 5 | 558 | 56 | ||
AK170546.1 | - | 4 - 5 | 726 | 56 | ||
BC137709.1 | - | 3 - 4 | 494 | 8860728 | 56 | |
BC137717.1 | - | 3 - 4 | 494 | 8860736 | 56 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AF288221.1 | - | 612 | Chr. - - [-] on Build | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |