Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300021715.mm.1 |
NM_001165965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021715.mm.1 |
NM_001165965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021715.mm.1 |
NM_177091.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021715.mm.1 |
NM_177091.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021715.mm.1 |
TC0300002755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021709.mm.1 |
NM_001165965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021709.mm.1 |
NM_001165965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021709.mm.1 |
NM_177091.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021709.mm.1 |
NM_177091.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021709.mm.1 |
TC0300002755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021714.mm.1 |
NM_001165965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021714.mm.1 |
NM_001165965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021714.mm.1 |
NM_177091.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021714.mm.1 |
NM_177091.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021714.mm.1 |
TC0300002755.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021713.mm.1 |
NM_001165965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021713.mm.1 |
NM_001165965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021713.mm.1 |
NM_177091.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021713.mm.1 |
NM_177091.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021713.mm.1 |
TC0300002755.mm.1 |
MTA TC |