Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026486.3 | NP_080762.1 | 14 - 15 | 1728 | 96 | |
GenBank mRNA | AK008223.1 | - | 3 - 4 | 252 | 96 | |
AK012720.1 | - | 14 - 15 | 1703 | 96 | ||
AK014476.1 | - | 14 - 15 | 1694 | 96 | ||
AK031829.1 | - | 14 - 15 | 1729 | 96 | ||
AK032378.1 | - | 13 - 14 | 1813 | 96 | ||
AK162182.1 | - | 1486 | 96 | |||
BC058375.1 | - | 14 - 15 | 1591 | 6828178 | 96 | |
DQ278869.1 | - | 14 - 15 | 1726 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |