Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026486.3 | NP_080762.1 | 11 - 12 | 1429 | 88 | |
GenBank mRNA | AK014476.1 | - | 11 - 12 | 1395 | 88 | |
AK031829.1 | - | 11 - 12 | 1430 | 88 | ||
AK032378.1 | - | 10 - 11 | 1514 | 88 | ||
AK162182.1 | - | 1187 | 88 | |||
BC058375.1 | - | 11 - 12 | 1292 | 6828178 | 88 | |
DQ278869.1 | - | 11 - 12 | 1427 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |