Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289472.1 | NP_001276401.1 | 13 - 14 | 2356 | 85 | |
NM_011256.3 | NP_035386.1 | 14 - 15 | 2547 | 85 | ||
GenBank mRNA | AF058693.1 | - | 13 - 14 | 2215 | 85 | |
AK129364.1 | - | 13 - 14 | 2356 | 85 | ||
BC082303.1 | - | 14 - 15 | 2547 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |