Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026831.1 | NP_081107.1 | 1 - 2 | 240 | 92 | |
GenBank mRNA | AK004148.1 | - | 1 - 2 | 241 | 92 | |
BC089625.1 | - | 1 - 2 | 228 | 6756382 | 92 | |
BC132636.1 | - | 1 - 2 | 229 | 40130913 | 92 | |
BC132638.1 | - | 1 - 2 | 229 | 40130915 | 92 | |
BY704393.1 | - | 1 - 2 | 241 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |