Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281999.1 | NP_001268928.1 | 11 - 12 | 1348 | 102 | |
NM_013832.4 | NP_038860.2 | 12 - 13 | 1324 | 102 | ||
GenBank mRNA | AF086714.1 | - | 11 - 12 | 1196 | 102 | |
AK156050.1 | - | 11 - 12 | 1312 | 102 | ||
BC005418.1 | - | 12 - 13 | 1338 | 3488024 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |