Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sort1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271599.1 | NP_001258528.1 | 16 - 17 | 2210 | 72 | |
NM_019972.3 | NP_064356.2 | 16 - 17 | 2210 | 72 | ||
GenBank mRNA | AF175279.1 | - | 16 - 17 | 2139 | 72 | |
AK147442.1 | - | 16 - 17 | 2193 | 72 | ||
BC034129.1 | - | 13 - 14 | 1643 | 72 | ||
BC056343.1 | - | 16 - 17 | 2142 | 5704346 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |