Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009125.2 | NP_033151.2 | 9 - 10 | 1581 | 74 | |
GenBank mRNA | AK135976.1 | - | 4 - 5 | 538 | 74 | |
U70670.1 | - | 9 - 10 | 1123 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |