Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144912.3 | NP_659161.2 | 1 - 2 | 62 | 74 | |
GenBank mRNA | AK136504.1 | - | 1 - 2 | 100 | 74 | |
BC021784.1 | - | 1 - 2 | 62 | 4162418 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |