Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 18 - 19 | 1979 | 90 | |
NM_001199310.1 | NP_001186239.1 | 18 - 19 | 1987 | 90 | ||
NM_010715.2 | NP_034845.2 | 18 - 19 | 1904 | 90 | ||
GenBank mRNA | AK153993.1 | - | 18 - 19 | 1866 | 90 | |
AK160208.1 | - | 3 - 4 | 273 | 90 | ||
AK161793.1 | - | 11 - 12 | 1062 | 90 | ||
AK168643.1 | - | 18 - 19 | 1983 | 90 | ||
BC028287.1 | - | 18 - 19 | 1829 | 4911261 | 90 | |
BC138541.1 | - | 18 - 19 | 1987 | 8861561 | 90 | |
BC138542.1 | - | 18 - 19 | 1987 | 8861562 | 90 | |
U04674.1 | - | 17 - 18 | 1772 | 90 | ||
U19604.1 | - | 18 - 19 | 2017 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |