Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 24 - 25 | 2647 | 87 | |
NM_001199310.1 | NP_001186239.1 | 24 - 25 | 2655 | 87 | ||
NM_010715.2 | NP_034845.2 | 24 - 25 | 2572 | 87 | ||
GenBank mRNA | AK153993.1 | - | 24 - 25 | 2534 | 87 | |
AK160208.1 | - | 9 - 10 | 941 | 87 | ||
AK168643.1 | - | 24 - 25 | 2651 | 87 | ||
BC028287.1 | - | 24 - 25 | 2497 | 4911261 | 87 | |
BC138541.1 | - | 24 - 25 | 2655 | 8861561 | 87 | |
BC138542.1 | - | 24 - 25 | 2655 | 8861562 | 87 | |
U04674.1 | - | 23 - 24 | 2440 | 87 | ||
U19604.1 | - | 24 - 25 | 2685 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |