Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029841.4 | NP_001025012.1 | 1 - 2 | 259 | 72 | |
GenBank mRNA | AK036167.1 | - | 1 - 2 | 260 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons |