Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025412.2 | NP_079688.2 | 2 - 3 | 248 | 75 | |
GenBank mRNA | AK002912.1 | - | 2 - 3 | 179 | 75 | |
AK004291.1 | - | 2 - 3 | 252 | 75 | ||
AK013214.1 | - | 2 - 3 | 174 | 75 | ||
BC026536.1 | - | 2 - 3 | 169 | 4987954 | 75 | |
CX212991.1 | - | 2 - 3 | 255 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |