Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081286.2 | NP_001074755.2 | 4 - 5 | 3984 | 76 | |
GenBank mRNA | AJ250768.1 | - | 4 - 5 | 3984 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |