Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026278.3 | NP_080554.1 | 6 - 7 | 789 | 86 | |
GenBank mRNA | AK006731.1 | - | 984 | 86 | ||
AK016626.1 | - | 794 | 86 | |||
BC030908.1 | - | 4 - 5 | 409 | 86 | ||
BC051060.1 | - | 2 - 3 | 184 | 86 | ||
BC051150.1 | - | 6 - 7 | 789 | 6772814 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |