Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026278.3 | NP_080554.1 | 7 - 8 | 1024 | 143 | |
GenBank mRNA | AK006731.1 | - | 1219 | 143 | ||
AK015594.1 | - | 8 - 9 | 1235 | 143 | ||
AK016626.1 | - | 1029 | 143 | |||
AK137605.1 | - | 2 - 3 | 330 | 143 | ||
BC030908.1 | - | 5 - 6 | 644 | 143 | ||
BC051060.1 | - | 3 - 4 | 419 | 143 | ||
BC051150.1 | - | 7 - 8 | 1024 | 6772814 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |