Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253754.1 | NP_001240683.1 | 2 - 3 | 406 | 73 | |
NM_153581.6 | NP_705809.1 | 2 - 3 | 777 | 73 | ||
GenBank mRNA | AK053538.1 | - | 2 - 3 | 331 | 73 | |
AK081877.1 | - | 2 - 3 | 663 | 73 | ||
AK147245.1 | - | 2 - 3 | 564 | 73 | ||
AK147280.1 | - | 2 - 3 | 564 | 73 | ||
BC023461.1 | - | 1 - 2 | 204 | 5041793 | 73 | |
BC024759.1 | - | 2 - 3 | 304 | 5364039 | 73 | |
BC029683.1 | - | 2 - 3 | 346 | 5365175 | 73 | |
BC029711.1 | - | 2 - 3 | 300 | 5358632 | 73 | |
BC033357.1 | - | 2 - 3 | 346 | 5363493 | 73 | |
BC033394.1 | - | 2 - 3 | 304 | 5364685 | 73 | |
BC043130.1 | - | 2 - 3 | 592 | 6810388 | 73 | |
CK623810.1 | - | 2 - 3 | 219 | 73 | ||
CX564488.1 | - | 1 - 2 | 198 | 73 | ||
S65735.1 | - | 2 - 3 | 303 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |