Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164607.1 | NP_001158079.1 | 6 - 7 | 965 | 77 | |
NM_001164608.1 | NP_001158080.1 | 5 - 6 | 872 | 77 | ||
NM_026859.3 | NP_081135.3 | 5 - 6 | 919 | 77 | ||
GenBank mRNA | AK004406.1 | - | 5 - 6 | 592 | 77 | |
AK042913.1 | - | 5 - 6 | 813 | 77 | ||
AK154047.1 | - | 5 - 6 | 862 | 77 | ||
AK189875.1 | - | 1 - 2 | 131 | 77 | ||
BC016260.1 | - | 5 - 6 | 907 | 4914167 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |