Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500004093.mm.1 |
NM_001164607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004093.mm.1 |
NM_001164607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004093.mm.1 |
NM_001164608.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004093.mm.1 |
NM_001164608.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004093.mm.1 |
NM_026859.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004093.mm.1 |
NM_026859.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004093.mm.1 |
TC1500000588.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004094.mm.1 |
NM_001164607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004094.mm.1 |
NM_001164607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004094.mm.1 |
NM_001164608.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004094.mm.1 |
NM_001164608.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004094.mm.1 |
NM_026859.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004094.mm.1 |
NM_026859.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004094.mm.1 |
TC1500000588.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004095.mm.1 |
NM_001164607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004095.mm.1 |
NM_001164607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004095.mm.1 |
NM_001164608.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004095.mm.1 |
NM_001164608.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004095.mm.1 |
NM_026859.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004095.mm.1 |
NM_026859.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004095.mm.1 |
TC1500000588.mm.1 |
MTA TC |