Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010232.1 | - | 9 - 10 | 828 | 73 | |
BC034895.1 | - | 8 - 9 | 607 | 73 | ||
BC061187.1 | - | 11 - 12 | 817 | 30277245 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |