Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164600.1 | NP_001158072.1 | 8 - 9 | 1013 | 145 | |
NM_028364.2 | NP_082640.2 | 9 - 10 | 1094 | 145 | ||
NM_133691.5 | NP_598452.2 | 8 - 9 | 1043 | 145 | ||
GenBank mRNA | AK075960.1 | - | 8 - 9 | 1011 | 145 | |
AK146534.1 | - | 9 - 10 | 1065 | 145 | ||
AK149632.1 | - | 9 - 10 | 1060 | 145 | ||
AK151526.1 | - | 8 - 9 | 1007 | 145 | ||
AK168789.1 | - | 8 - 9 | 1010 | 145 | ||
BC010601.1 | - | 8 - 9 | 1013 | 3257153 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |