Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159321.1 | NP_001152793.1 | 4 - 5 | 1100 | 60 | |
NM_133355.1 | NP_579933.1 | 3 - 4 | 404 | 60 | ||
GenBank mRNA | AF099933.2 | - | 3 - 4 | 404 | 60 | |
BC075624.1 | - | 456 | 6841572 | 60 | ||
DQ193535.1 | - | 4 - 5 | 1100 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |