Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900004407.mm.1 |
NM_029682.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004407.mm.1 |
NM_029682.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004407.mm.1 |
TC1900000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004417.mm.1 |
NM_029682.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004417.mm.1 |
NM_029682.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004417.mm.1 |
TC1900000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004409.mm.1 |
NM_029682.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004409.mm.1 |
NM_029682.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004409.mm.1 |
TC1900000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004411.mm.1 |
NM_029682.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004411.mm.1 |
NM_029682.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004411.mm.1 |
TC1900000495.mm.1 |
MTA TC |