Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021403.3 | NP_067378.2 | 3 - 4 | 861 | 86 | |
GenBank mRNA | BC051947.1 | - | 3 - 4 | 468 | 5696610 | 86 |
BC058385.1 | - | 3 - 4 | 861 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |