Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177369.1 | NP_001170840.1 | 20 - 21 | 2453 | 110 | |
NM_001177370.1 | NP_001170841.1 | 19 - 20 | 2369 | 110 | ||
NM_019408.3 | NP_062281.1 | 19 - 20 | 2604 | 110 | ||
GenBank mRNA | AF155372.1 | - | 20 - 21 | 2531 | 110 | |
AF155373.1 | - | 20 - 21 | 2568 | 110 | ||
AK134081.1 | - | 20 - 21 | 2872 | 110 | ||
AK137680.1 | - | 20 - 21 | 2443 | 110 | ||
AK144994.1 | - | 20 - 21 | 2446 | 110 | ||
AK145312.1 | - | 19 - 20 | 2570 | 110 | ||
AK148160.1 | - | 20 - 21 | 2441 | 110 | ||
AK150012.1 | - | 20 - 21 | 2432 | 110 | ||
AK153412.1 | - | 15 - 16 | 2013 | 110 | ||
AK181809.1 | - | 283 | 110 | |||
BC027423.1 | - | 19 - 20 | 2370 | 3672913 | 110 | |
DQ020183.1 | - | 18 - 19 | 2422 | 110 | ||
DQ020185.1 | - | 19 - 20 | 2301 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |