Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133895.1 | NP_598656.1 | 5 - 6 | 1281 | 99 | |
GenBank mRNA | AK048529.1 | - | 5 - 6 | 1248 | 99 | |
AK154238.1 | - | 5 - 6 | 98 | |||
BC016233.1 | - | 5 - 6 | 1281 | 4502184 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |