Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008806.2 | NP_032832.2 | 11 - 12 | 1532 | 59 | |
GenBank mRNA | AK039397.1 | - | 11 - 12 | 1244 | 59 | |
AK044364.1 | - | 11 - 12 | 1475 | 59 | ||
BC048247.1 | - | 11 - 12 | 1511 | 59 | ||
BC058803.1 | - | 10 - 11 | 2192 | 59 | ||
BC129923.1 | - | 11 - 12 | 1462 | 40111882 | 59 | |
BC129924.1 | - | 11 - 12 | 1461 | 59 | ||
X55968.1 | - | 11 - 12 | 1465 | 59 | ||
X60133.1 | - | 11 - 12 | 1472 | 59 | ||
X87952.1 | - | 11 - 12 | 1472 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |