Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008806.2 | NP_032832.2 | 19 - 20 | 2336 | 66 | |
GenBank mRNA | AK039397.1 | - | 19 - 20 | 2048 | 66 | |
AK044364.1 | - | 19 - 20 | 2279 | 66 | ||
BC048247.1 | - | 19 - 20 | 2315 | 66 | ||
BC058803.1 | - | 15 - 16 | 3683 | 66 | ||
BC129923.1 | - | 19 - 20 | 2266 | 40111882 | 66 | |
BC129924.1 | - | 19 - 20 | 2265 | 66 | ||
X60133.1 | - | 19 - 20 | 2276 | 66 | ||
X87952.1 | - | 19 - 20 | 2276 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |