Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148935.2 | NP_683737.2 | 2 - 3 | 160 | 138 | |
GenBank mRNA | AF323488.1 | - | 2 - 3 | 168 | 138 | |
AY039039.1 | - | 2 - 3 | 206 | 138 | ||
BC092242.1 | - | 2 - 3 | 149 | 30698162 | 138 | |
BC130222.1 | - | 2 - 3 | 161 | 40141600 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |