Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023742.2 | NP_076231.1 | 6 - 7 | 1554 | 102 | |
GenBank mRNA | AB015423.1 | - | 6 - 7 | 1477 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |