Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178398.4 | NP_848485.1 | 4 - 5 | 601 | 99 | |
GenBank mRNA | AK010881.1 | - | 4 - 5 | 629 | 99 | |
AK155113.1 | - | 4 - 5 | 631 | 99 | ||
AK169051.1 | - | 4 - 5 | 575 | 99 | ||
AK172030.1 | - | 4 - 5 | 577 | 99 | ||
BC044894.1 | - | 4 - 5 | 626 | 4235237 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |