Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080808.1 | NP_001074277.1 | 5 - 6 | 1100 | 99 | |
GenBank mRNA | AK036757.1 | - | 2 - 3 | 560 | 99 | |
BC024804.1 | - | 5 - 6 | 1093 | 99 | ||
BC126948.1 | - | 5 - 6 | 1100 | 40090576 | 99 | |
GU973876.1 | - | 5 - 6 | 1100 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |